Epic High Performance Static Dissipative Frocks | Avantor
Epic High Performance Static Dissipative Frocks
White, 5XL
Catalog # 77394-786
Fournisseur #:  846855-5XL
Epic High Performance Static Dissipative Frocks
Catalog # 77394-786
Epic High Performance Static Dissipative Frocks
Epic High Performance Static Dissipative Frocks
White, 5XL
Price
$171.51
Log in pour la disponibilité
Case of 30
Product Details & Documents
Lint-free film coated frock for use in critical, ESD and low particulate clean environments. Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas.
  • Snap front closures
  • Knit wrists
  • Knit collar
  • No pockets
Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane. The finished fabric contains a proprietary additive making it static dissipative. Individually inspected for sizing, stitching and workmanship.
White garment with white snaps, white knit wrists, and white knit collar.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case.
Spécifications
  • Taille de confection:
    5XL
  • Color:
    White
  • Cat. No.:
    77394-786
  • Sleeve or leg length:
    Long
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps