Epic High Performance Static Dissipative Frocks
Product Details & Documents
Lint-free film coated frock for use in critical, ESD and low particulate clean environments. Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas.
- Snap front closures
- Knit wrists
- Knit collar
- No pockets
Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane. The finished fabric contains a proprietary additive making it static dissipative. Individually inspected for sizing, stitching and workmanship.
White garment with white snaps, white knit wrists, and white knit collar.
Packaging: Neatly folded and cleanroom packed; 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case.
Spécifications
- Taille de confection:3XL
- Color:White
- Cat. No.:77394-782
- Sleeve or leg length:Long
- Pockets:Without pockets
- Closure type:Press studs / snaps