5×3 X-Gen Spectacles | Avantor
5×3 X-Gen Spectacles
Scratch-resistant/hardcoat, Clear, Black/green
Catalog # 77393-750
Fournisseur #:  MDU5031SC
5×3 X-Gen Spectacles
Catalog # 77393-750
5×3 X-Gen Spectacles
5×3 X-Gen Spectacles
Scratch-resistant/hardcoat, Clear, Black/green
Price
$198.03
Log in pour la disponibilité
Pack of 10
Product Details & Documents
The 5×3 spectacles feature floating lens frame technology, which adapts to face conformation with no need for adjustments.
  • Ultra-wraparound lens for better protection around the eyes
  • Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
Designed to avoid annoying contact points with the face, these spectacles feature a frame with Floating Lens technology, available with anti-fog and anti-scratch ultra performing vanguard ultra (UDC) treatment. UV525 treatment is also available to protect eyes during curing light usage.
Spécifications
  • Frame Color:
    Black/green
  • Lens Coating:
    Scratch-resistant/hardcoat
  • Lens Tint:
    Clear