Face Shields, 711
Replacement shields, Clear
Catalog # 77393-784
Supplier: UNIVET OPTICAL TECHNOLOGIES
Supplier #: MDU7019S
Jump to:
Product Details & Documents
The 711 series face shields feature an anti-fog lens treatment.
- Anti-fog lens treatment
- Disposable and interchangeable shields
- Ultra-lightweight design
- Multiple frame colors available
The 711 series face shields can be worn with prescription glasses and protective masks.
Ordering information: Anti-fog replacement shields are available separately.
Spécifications
- Désignation:Replacement shields
- Lens Tint:Clear