Face Shield, 701.02
Replacement face shield, Clear
Catalog # 77393-782
Supplier: UNIVET OPTICAL TECHNOLOGIES
Fournisseur #: MDU7012S
Jump to:
Product Details & Documents
The 701.02 series face shield is made from clear acetate, and features an anti-fog lens treatment.
- Anti-fog acetate shield
- Adjustable elastic head band
- Shield is inclinable up to 90°
- Clear/gray frame, clear lens
Designed with an interchangeable shield, the 701.02 series face shield can be worn with prescription glasses and protective masks. This protective device is certified according to EN 166, level 3 protection (EU) against droplets and splashes of liquids.
Ordering information: Anti-fog acetate replacement shields are available separately.
Spécifications
- Désignation:Replacement face shield
- Lens Tint:Clear