Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
HighSpec Rho1D4 Agarose

HighSpec Rho1D4 Agarose

50 ml
Catalog # 77393-066
Supplier:   Cube Biotech
Supplier #:  33103
HighSpec Rho1D4 Agarose
Catalog # 77393-066
Supplier:  Cube Biotech
CAS Number:  
HighSpec Rho1D4 Agarose
HighSpec Rho1D4 Agarose

50 ml

CAS Number:  
Price
$3,858.20
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

HighSpec Rho1D4 Agarose resin is a suspension of 40 µm agarose beads, coupled to an anti-Rho1D4 antibody. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead Ligand: anti-Rho1D4 antibody
  • Bead size: 100 μm
  • Matrix: 6% cross-linked agarose
  • Recommended flow rates: 0.25 to 1 ml/min
  • Max. flow rates: 6 ml/min
  • Filling quantity: Delivered as a 50% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C
  • Required equipment:
  • Lysis Buffer
  • Wash Buffer
  • Elution Buffer (incl. Rho1D4 peptide)
  • Ice bath
  • Refrigerated centrifuge for 50 ml tube (min 10000×g)
  • 50 ml centrifuge tube
  • Micropipettor and Micropipetting tips
  • Disposable gravity flow columns with capped bottom outlet, 2 ml
  • pH meter
  • End-over-end shaker
  • SDS-PAGE buffers, reagents and equipment Optional: Western Blot reagents and equipment

Protein affinity epitope tags find use in numerous applications in life sciences. Ranging from Western Blot detection, protein localization, ELISA assays, and most importantly protein purification. The possible uses for epitope tags are numerous (Molday and Molday 2014). Rho1D4, sometimes also just called '1D4-tag', is an epitope tag that originates from the C-terminal end of bovine rhodopsin. Its nine amino acids long sequence is TETSQVAPA, which gives it a size of 902.9 Dalton. The affinity tag and the specific monoclonal antibodies were discovered in 1984 by Hodges et al. and MacKenzie et al.

Specifications

Specifications

  • Taille:
    50 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.