Face Shields, 711
711 face shield, Pink, Clear
Catalog # 77393-776
Supplier: UNIVET OPTICAL TECHNOLOGIES
: MDU7007S
Jump to:
Product Details & Documents
The 711 series face shields feature an anti-fog lens treatment.
- Anti-fog lens treatment
- Disposable and interchangeable shields
- Ultra-lightweight design
- Multiple frame colors available
The 711 series face shields can be worn with prescription glasses and protective masks.
: Anti-fog replacement shields are available separately.
Spécifications
- Frame Color:Pink
- Désignation:711 face shield
- Lens Tint:Clear