Face Shield, 701.02
701.02 face shield, Clear
Catalog # 77393-772
Supplier: UNIVET OPTICAL TECHNOLOGIES
: MDU7002S
Jump to:
Product Details & Documents
The 701.02 series face shield is made from clear acetate, and features an anti-fog lens treatment.
- Anti-fog acetate shield
- Adjustable elastic head band
- Shield is inclinable up to 90°
- Clear/gray frame, clear lens
Designed with an interchangeable shield, the 701.02 series face shield can be worn with prescription glasses and protective masks. This protective device is certified according to EN 166, level 3 protection (EU) against droplets and splashes of liquids.
: Anti-fog acetate replacement shields are available separately.
Spécifications
- Désignation:701.02 face shield
- Lens Tint:Clear