546 Value Spectacles
Anti-fog, scratch-resistant, Clear, Black/green
Catalog # 77393-762
Supplier: UNIVET OPTICAL TECHNOLOGIES
: MDU5038SC
Jump to:
Product Details & Documents
Extremely lightweight and modern spectacles with a sporty line.
- Multiple lens treatments are available, including anti-fog, anti-scratch, and UV525
- Wraparound polycarbonate lens
- Integrated side shields
546 Value spectacles feature temples equipped with rubber terminals to guarantee excellent comfort, as well as a non-slip rubber nose pad. The UV525 protects eyes during curing light usage. Smoke lenses can help to reduce anxiety.
Spécifications
- Frame Color:Black/green
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear