Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

PureCube NTA Agarose

50 ml
Catalog # 77393-010
 :  31705
PureCube NTA Agarose
Catalog # 77393-010
Supplier:  
CAS Number:  
PureCube NTA Agarose
PureCube NTA Agarose

50 ml

CAS Number:  
Discontinued
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

PureCube NTA (nitrilotriacetic acid) agarose resin was developed for IMAC-based protein purification and protein enrichment methods.

  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: NTA
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

The beads in this suspension have an average diameter of 40 µm and, thus, excellent mechanical stability. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA agarose resin uncoupled to any metal ion in case we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Spécifications

Spécifications

  • Taille:
    50 ml

Spécifications