Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PureCube NTA Agarose | Avantor

PureCube NTA Agarose

1 ml
Catalog # 77393-006
PureCube NTA Agarose
Catalog # 77393-006
Supplier:  
CAS Number:  
PureCube NTA Agarose
PureCube NTA Agarose

1 ml

CAS Number:  
Discontinued
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

PureCube NTA (nitrilotriacetic acid) agarose resin was developed for IMAC-based protein purification and protein enrichment methods.

  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: NTA
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

The beads in this suspension have an average diameter of 40 µm and, thus, excellent mechanical stability. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA agarose resin uncoupled to any metal ion in case we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Spécifications

Spécifications

  • Taille:
    1 ml

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.