Ward's® Goethite
Ward's® Goethite
Goethite, student specimens, 1×1 - 1×1"
Catalog # 470354-424
Supplier:   Avantor
Ward's® Goethite
Catalog # 470354-424
Supplier:  Avantor
Ward's® Goethite
Goethite, student specimens, 1×1 - 1×1"
Price
$23.09
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Iron oxide-hydroxide mineral; reddish brown to black color; crystal habit ranges from massive to acicular.
  • Collected in Michigan
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage boxes (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Spécifications
  • Désignation:
    Goethite, student specimens
  • Specimen Dimensions:
    1×1 - 1×1"