WHEATON® Serum Vials
Serum vial, 10 ml, USBB, clear, EP, case, 10 ml (brimful capacity), 22×40 mm, Glass, Clear, Non
Catalog # 77392-014
Supplier: DWK Life Sciences (KIMBLE)
: W0EBB06R-C-EP
Jump to:
Product Details & Documents
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Spécifications
- Avec bouchon:Non
- Capacité:10 ml (brimful capacity)
- Color:Clear
- Désignation:Serum vial, 10 ml, USBB, clear, EP, case
- Matière:Glass
- Diameter ×H:22×40 mm



