WHEATON® Serum Vials
Serum vial, 4 ml, USBB, clear, EP, pallet, 4 ml (brimful capacity), 16×35 mm, Glass, Clear, Non
77392-024
: W0EBB02R-P-EP
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
- Avec bouchon:Non
- Capacité:4 ml (brimful capacity)
- Color:Clear
- Désignation:Serum vial, 4 ml, USBB, clear, EP, pallet
- Matière:Glass
- Diameter ×H:16×35 mm



