Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
WHEATON® Serum Vials | Avantor

WHEATON® Serum Vials

Serum vial, 4 ml, USBB, clear, EP, case, 4 ml (brimful capacity), 16×35 mm, Glass, Clear, Non
Catalog # 77392-010
Fournisseur #:  W0EBB02R-C-EP
WHEATON® Serum Vials
Catalog # 77392-010
WHEATON® Serum Vials
WHEATON® Serum Vials

Serum vial, 4 ml, USBB, clear, EP, case, 4 ml (brimful capacity), 16×35 mm, Glass, Clear, Non

Price
$2,165.80
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 2,352
Product Details & Documents

Serum vials converted from Type I borosilicate glass tubing.

  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
  • Crimp cap type

Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.

Spécifications
  • Avec bouchon:
    Non
  • Capacité:
    4 ml (brimful capacity)
  • Color:
    Clear
  • Désignation:
    Serum vial, 4 ml, USBB, clear, EP, case
  • Matière:
    Glass
  • Diameter ×H:
    16×35 mm