WHEATON® Serum Vials
Serum vial, 5 ml, USBB, clear, EP, case, 5 ml, 21.75×40 mm, Glass, Clear, Non
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: W0UBB5MLC-C-EP
Jump to:
Product Details & Documents
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
Specifications
- Avec bouchon:Non
- Capacité:5 ml
- Color:Clear
- Désignation:Serum vial, 5 ml, USBB, clear, EP, case
- Matière:Glass
- Diameter ×H:21.75×40 mm



