QuickConnect™ Mop Handles
QuickConnect™ carbon fiber telescopic handle, 175 - 848 cm (69 - 334")
Catalog # 77322-294
Supplier: Contec
Fournisseur #: CFH334
Jump to:
Product Details & Documents
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
- Compatible with all QuickConnect™ frames
- Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
Documents
Spécifications
- Désignation:QuickConnect™ carbon fiber telescopic handle
- Longueur:175 - 848 cm (69 - 334")