Raspberry Pi 4 Computers | Avantor
Raspberry Pi 4 Computers
Raspberry Pi 4 SBC, 8 GB RAM
Catalog # 470354-022
Supplier:   NEWARK ELECTRONICS
Fournisseur #:  64AH2041
Raspberry Pi 4 Computers
Catalog # 470354-022
Supplier:  NEWARK ELECTRONICS
Raspberry Pi 4 Computers
Raspberry Pi 4 Computers
Raspberry Pi 4 SBC, 8 GB RAM
Price
$200.39
Log in pour la disponibilité
Each
Product Details & Documents
Ground-breaking increases in speed and performance compared to the Raspberry Pi 3 Model B+.
  • Choose either 4 or 8 GB of RAM
  • Single board computer only; power adapter, accessories, display cable, and microSD card available separately
For the end user, Raspberry Pi 4 Model B provides desktop performance comparable to entry-level x86 PC systems. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
Spécifications
  • Désignation:
    Raspberry Pi 4 SBC, 8 GB RAM