Maintenance Solutions for Electrodes
Maintenance Solutions for Electrodes
InLab storage solution, 250 ml
Catalog # 77322-214
Supplier:   Mettler Toledo
Supplier #:  30111142
Maintenance Solutions for Electrodes
Catalog # 77322-214
Supplier:  Mettler Toledo
CAS Number:  
Maintenance Solutions for Electrodes
Maintenance Solutions for Electrodes
InLab storage solution, 250 ml
CAS Number:  
Price
$64.30
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This solution keeps your electrode ready with the right cleaning, storage and regeneration solution.
Delivery information: Includes quality inspection certificate.
Packaging: Packaging type - bottle.
Specifications
Specifications
  • Désignation:
    InLab storage solution
  • Taille:
    250 ml