Screen Sieve Set 6
Screen sieve, 230
470353-768
: SSC-15-230
An economical set of durable sieve that will hold up to tough use.
- Set includes 6 popular mesh sizes
- The frame is made from durable steel
- A top and bottom cover are provided for storage
- Each screan is labeled with the mesh size
- 15 cm diameter for all sieves
The process of screening allows students to study the make up of the materials they have collected. The screening process athat aid in the recognition of the different parts of a specimen. The screen sieves are a valuable tool used in this process. This set features steel screens with six different sizes of mess that include: 5, 10, 35, 60, 120, 230. A top and bottom is also provided to enclose the set.
: Each screen can be purchased individual.
- Désignation:Screen sieve
- Mesh Size:230