FILTER POLYFLOW G 1UM EPDM O-RING 30INCH
77320-724
Frequently Bought Together
VWR® Specimen Containers with Dual Click
: VWR International
Glycérol ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
: VWR International
VWR® Multipurpose Containers, HDPE
: VWR International
COD Digestion vials, VWR Chemicals BDH®
: VWR International
Histoplex™ Histology Containers, Starplex®
: Starplex
VACUETTE® Secondary Tubes, Greiner Bio-One
: Greiner Bio-One
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
: TCI America
Corning® Dispase
: Corning
SingleQuots® EGM®-2 Endothelial Kit
: LONZA PHARMA - BIOSCIENCE CA
Solvent Resistant Pen, Electron Microscopy Sciences
: Electron Microscopy Sciences
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
: Sekisui Diagnostics
Ether de pétrole, 35…60 °C ACS
: AVANTOR PERFORMANCE MATERIAL LLC
TapeStation Systems Parts and Accessories
: AGILENT TECHNOLOGIES (GENOMICS) CA