Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
J.T.Baker® Cell Lysis Solution, Biotech Reagent

J.T.Baker® Cell Lysis Solution, Biotech Reagent

Cell Lysis Solution, Biotech Reagent GMP, 5000 ml, Plastic bottle
Catalog # CAJTG193-09
   Safer Human and Environmental Health
Supplier #:  BAKRG193-09
J.T.Baker® Cell Lysis Solution, Biotech Reagent
Catalog # CAJTG193-09
   Safer Human and Environmental Health
CAS Number:  
J.T.Baker® Cell Lysis Solution, Biotech Reagent
J.T.Baker® Cell Lysis Solution, Biotech Reagent

Cell Lysis Solution, Biotech Reagent GMP, 5000 ml, Plastic bottle

CAS Number:  
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each (5,000ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

J.T.Baker® Cell Lysis Solution Biotech Reagent is a cGMP pre-formulated and ready to use detergent optimized for the extraction and recovery of AAV particles from producer cell lines (HEK293 parental and suspension lines, SF9). J.T.Baker® Cell Lysis Solution is a proprietary blend of nonionic surfactants, free of octyl phenol ethoxylate, which demonstrates comparable performance industry standard detergents while addressing environmental concerns associated with REACH and downstream clearance concerns. Designed for Gene Therapy workflows, this formulation is easy to use and handle as it is ready-to-use, has low viscosity, does not require cold temperature shipping or storage, and is available in sizes to support processes from development to large scale commercial batches. Detail description provided in technical literature.

  • Comparable cell lysis performance to octyl phenol ethoxylate
  • Improved protection of viral particles
  • Easy to use and handle due to reduced foaming and viscosity and Room Termperature shipping and storage
  • 0.1 µm filtered solution

Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)

Chemical Specifications of GMP Cell lysis solution:
GMP Manufactured Product
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH : For information only
Viscosity (25°C) : For information only
Density: 0.9 – 1.2 g/mL
Total Aerobic Microbial Count: 100 cfu/mL
Total Yeast and Mold Count: 10 cfu/mL
UV-Vis (max Absorption) at 380-450 nm: For information only

Spécifications

Spécifications

  • Type de conditionnement:
    Plastic bottle
  • Désignation:
    Cell Lysis Solution, Biotech Reagent GMP
  • Taille:
    5000 ml
  • Cat. No.:
    CAJTG193-09
  • Environmental certification:
    EPA safer choice chemical
  • Environmentally Preferable:
    Safer Human and Environmental Health

Spécifications

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.