Code Corporation Barcode Reader Accessories
Scanner stand, black, 9×4.25×6", CR1500 (176510), CR1500 kit (176511), CR1100 (176697), CR1100 kit (176698), CR950 (176836)
  77282-360
 :  176856
Code Corporation Barcode Reader Accessories
  77282-360
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories
Scanner stand, black, 9×4.25×6", CR1500 (176510), CR1500 kit (176511), CR1100 (176697), CR1100 kit (176698), CR950 (176836)
$87.45
Each
Accessories available for the barcode readers.
Use the 6′ (176701) or 8' (176700) Coiled RS232 Affinity® Cable CRA-C511 with Code's cabled barcode readers.
Model 176702 Code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3.3 A).
Model 176703 Code power supply is an international power supply with USB connection.
Model 176856 - Use this stand to scan hands-free or simply to hold your scanner. It features a flexible gooseneck design that adjusts easily to different sized scanners.
  • Désignation:
    Scanner stand, black, 9×4.25×6"
  • For Model:
    CR1500 (176510), CR1500 kit (176511), CR1100 (176697), CR1100 kit (176698), CR950 (176836)