Code Corporation Barcode Reader Accessories
Power supply, black, 1.5×3×2.5", 19.7' cable length, CR2700 Quad bay battery charger (176520)
  77282-172
 :  176702
Code Corporation Barcode Reader Accessories
  77282-172
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories
Power supply, black, 1.5×3×2.5", 19.7' cable length, CR2700 Quad bay battery charger (176520)
$76.53
Each
Accessories available for the barcode readers.
Use the 6′ (176701) or 8' (176700) Coiled RS232 Affinity® Cable CRA-C511 with Code's cabled barcode readers.
Model 176702 Code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3.3 A).
Model 176703 Code power supply is an international power supply with USB connection.
Model 176856 - Use this stand to scan hands-free or simply to hold your scanner. It features a flexible gooseneck design that adjusts easily to different sized scanners.
  • Désignation:
    Power supply, black, 1.5×3×2.5", 19.7' cable length
  • For Model:
    CR2700 Quad bay battery charger (176520)