The Frog Microslides | Avantor
The Frog Microslides
The frog microslides, set of 15
Catalog # 470353-164
Fournisseur #:  T272-15
The Frog Microslides
Catalog # 470353-164
The Frog Microslides
The Frog Microslides
The frog microslides, set of 15
Price
$283.45
Log in pour la disponibilité
Box of 15
Product Details & Documents
Explore the tissues and organs of this model organism.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1.Frog skin (160×) 2. Frog gills 3. Frog lungs - L.S. stained (1×) 4. Frog blood (400×)/Human blood (900×) 5. Frog heart left- W.M. (2×)/right- L.S. (5×) L 6. Frog intestine - X.S. (20×) 7. Frog cloaca (2×) 8. Frog brain W.M. (2×).
Spécifications
  • Désignation:
    The frog microslides, set of 15
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.