The Frog Microslides | Avantor
The Frog Microslides
The frog microslides
Catalog # 470353-198
Fournisseur #:  T272
The Frog Microslides
Catalog # 470353-198
The Frog Microslides
The Frog Microslides
The frog microslides
Price
$19.51
Log in pour la disponibilité
Each
Product Details & Documents
Explore the tissues and organs of this model organism.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1.Frog skin (160×) 2. Frog gills 3. Frog lungs - L.S. stained (1×) 4. Frog blood (400×)/Human blood (900×) 5. Frog heart left- W.M. (2×)/right- L.S. (5×) L 6. Frog intestine - X.S. (20×) 7. Frog cloaca (2×) 8. Frog brain W.M. (2×).
Spécifications
  • Désignation:
    The frog microslides