Helpful and Harmful Bacteria Microslides
Helpful and Harmful Bacteria Microslides
Harmful and helpful bacteria microslides
Catalog # 470353-048
Supplier #:  T105
Helpful and Harmful Bacteria Microslides
Catalog # 470353-048
Helpful and Harmful Bacteria Microslides
Helpful and Harmful Bacteria Microslides
Harmful and helpful bacteria microslides
Price
19,51 $
Log in pour la disponibilité
Each
Product Details & Documents
Explore examples of both harmful and helpful bacteria.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Three-types of bacteria of Diphtheria - stained (2000×) 2. Bacteria of Diphtheria - stained - (2000×) 3. Bacteria of typhoid fever - stained (2000×) 4. Bacteria of food poisoning - stained -(2000) 5. Bacteria of potato ring rot - stained (2000×) 6. Bacteria of blood poisoning - stained - (2000×) 7. Cheese bacteria - stained - (2000) 8. Vinegar bacteria - stained - (2000×).
Spécifications
  • Désignation:
    Harmful and helpful bacteria microslides