Taxonomy Microslides | Avantor
Taxonomy Microslides
Taxonomy microslides, set of 15
Catalog # 470353-134
Fournisseur #:  T217-15
Taxonomy Microslides
Catalog # 470353-134
Taxonomy Microslides
Taxonomy Microslides
Taxonomy microslides, set of 15
Price
$283.45
Log in pour la disponibilité
Box of 15
Product Details & Documents
Explore the classification systems of organisms.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Bacteria in pairs - (1200×) 2. A spore producer - stained - (1000×) 3. Bacteria in chains - stained - (1000×) 4. Rod shaped bacteria - stained - (2000×) 5. Another spore producer - stained - (1000×) 6. Boils and abscess bacteria - stained - (1000×) 7. Syphilis bacterium - electron micrograph - (30000×) 8. Sauerkraut bacteria - stained - (2000×).
Spécifications
  • Désignation:
    Taxonomy microslides, set of 15
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.