Taxonomy Microslides
Product Details & Documents
Explore the classification systems of organisms.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson pan on the topic under study
- Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Bacteria in pairs - (1200×) 2. A spore producer - stained - (1000×) 3. Bacteria in chains - stained - (1000×) 4. Rod shaped bacteria - stained - (2000×) 5. Another spore producer - stained - (1000×) 6. Boils and abscess bacteria - stained - (1000×) 7. Syphilis bacterium - electron micrograph - (30000×) 8. Sauerkraut bacteria - stained - (2000×).
Spécifications
- Désignation:Taxonomy microslides, set of 15