Fossils and Evolution Microslides
Product Details & Documents
Explore evolution as it presents in the fossil record over time.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson pan on the topic under study
- Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
: Viewers are available separately.
: Set includes 1. Fossil starfish (Mesopalaester) (1/2×) 2. Eospirifer (left side 2×), Microspirifer (right side 1.5×) 3. Jurassic Spirifer (2×) 4. Onoceras (1×) 5. Hoploscapites (1/3×) 6. Heohippus skull ( 1/5×) 7. Equus teeth (1/2×) 8. Primate feet (2×).
Spécifications
- Désignation:Fossils and evolution microslides