Introduction to Space Microslides | Avantor
Introduction to Space Microslides
Introduction to space microslides, set of 15
Catalog # 470353-084
Fournisseur #:  T130-15
Introduction to Space Microslides
Catalog # 470353-084
Introduction to Space Microslides
Introduction to Space Microslides
Introduction to space microslides, set of 15
Price
$464.65
Log in pour la disponibilité
Box of 15 Case of 150
Product Details & Documents
One strip of 8 related 35 mm images photographed through a microscope called photomicrographs.
Arrows and call outs help the student locate important features. The film is mounted in a clear plastic holder that protects it on both sides.
Each microslide strip contains the following photomicrographs:
1. The Moon
2. The Sun
3. Planets
4. Meteorodis
5. Comets
6. Neighboring Stars
7. Our Galaxy
8. Other Galaxies
Each Microslide® is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the Microslide® is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Spécifications
  • Désignation:
    Introduction to space microslides, set of 15
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.