Eisco Plastic Scoops
Plastic scoops, 1000 ml
Catalog # 470345-467
Supplier: Wards
Supplier #: CH0461IPK6
Jump to:
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Spécifications
- Capacité:1000 ml
- Désignation:Plastic scoops