Eisco Plastic Scoops
Plastic scoops, 250 ml
Catalog # 470346-053
Supplier: Wards
Fournisseur #: CH0461GPK6
Jump to:
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Spécifications
- Capacité:250 ml
- Désignation:Plastic scoops
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.