Eisco Plastic Scoops | Avantor
Eisco Plastic Scoops
Plastic scoops, 250 ml
Catalog # 470346-053
Supplier:   Wards
Fournisseur #:  CH0461GPK6
Eisco Plastic Scoops
Catalog # 470346-053
Supplier:  Wards
Eisco Plastic Scoops
Eisco Plastic Scoops
Plastic scoops, 250 ml
Price
$45.10
Log in pour la disponibilité
Pack of 6
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
  • Made of heat- and chemical-resistant polypropylene plastic
  • Scoop handles are labeled with capacity for easy identification
  • Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Spécifications
  • Capacité:
    250 ml
  • Désignation:
    Plastic scoops