BoneClones® Replica Teeth
African elephant tooth, 1, 30.4 cm
Catalog # 470352-908
Supplier: BONE CLONES HOLDINGS
Fournisseur #: KO-045
Jump to:
Product Details & Documents
Compare tooth shape based on diet.
- Explore the influences of diet and use in tooth shape
- Highly detailed replicas from real specimens
Teeth serve many different functions in the animal world, each of the teeth below have evolved based on carnivorous nature or the animal. What does the structure of each tell about the feeding habits of the animal?
Spécifications
- Désignation:African elephant tooth
- Longueur:30.4 cm
- No. of Parts:1