LabFit™ Base Cabinet Sinks
Catalog # 77135-370
Supplier:   TrippNT
 :  53696
LabFit™ Base Cabinet Sinks
Catalog # 77135-370
Supplier:  TrippNT
LabFit™ Base Cabinet Sinks
LabFit™ Base Cabinet Sinks
Price
$3,976.65
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Embedded magnetic door latches
  • Large removeable rear access panel
  • Sink not included
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistnace to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery information: TrippNT® LabFit™ laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.