Hooks Collars
Hooks collar, Plastic, 19 mm Int.Ø
Catalog # 470352-506
Supplier: GSC International
Supplier #: HKCLR-19-C
Jump to:
Product Details & Documents
The Hooks Collar is constructed with a ring that fits over a support stand rod, a hook and thumb screw.
- Metal ring
- Metal hook
- Different sizes and materials to fit all classroom needs and budgets
The Hooks Collar is used in a physics classroom for suspending items such as pulleys, cords, or weights from a rod or crass bar.
Caution: Prop 65 Lead Warnings are attached to the package.
Specifications
- Désignation:Hooks collar
- Matière:Plastic
- Taille:19 mm Int.Ø