Hooks Collars
Hooks Collars
Hooks collar, Plastic, 13 mm Int.Ø
Catalog # 470352-508
Supplier:   GSC International
Supplier #:  HKCLR-13-C
Hooks Collars
Catalog # 470352-508
Supplier:  GSC International
Hooks Collars
Hooks Collars
Hooks collar, Plastic, 13 mm Int.Ø
Price
$5.70
Log in pour la disponibilité
Each
Product Details & Documents
The Hooks Collar is constructed with a ring that fits over a support stand rod, a hook and thumb screw.
  • Metal ring
  • Metal hook
  • Different sizes and materials to fit all classroom needs and budgets
The Hooks Collar is used in a physics classroom for suspending items such as pulleys, cords, or weights from a rod or crass bar.
Caution: Prop 65 Lead Warnings are attached to the package.
Spécifications
  • Désignation:
    Hooks collar
  • Matière:
    Plastic
  • Taille:
    13 mm Int.Ø