SevenDirect™ SD20 pH Meter
Product Details & Documents
The pH meter SevenDirect™ SD20 provides accurate pH measurements for almost any sample across a wide range of applications. The EasyPlace™ electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
- Intuitive user interface using 7" touchscreen
- On-screen instructions guide through the calibration process
- EasyPlace™ electrode arm ensures precise placement of the sensor
- Feature a closed connector compartment to prevent corrosion
- A replacable protective cover keeps the instrument safe from dust and spills (IP 54 in-use)
- Instruments support GLP compliance by storing key measurement data
The intuitive user interface of the 7" touchscreen facilitates the measurement process by avoiding unnecessary steps and focusing on the most important information. The SevenDirect™ has a guided verification process that provides a straightforward way to check system performance with a clear pass/fail indication. The meter has a replaceable protective cover to keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use).
Delivery information: The meter-only model includes EasyPlace™ electrode holder, protective cover, Get-Started guide and power supply. The kits also include an InLab sensor and 2 buffer sachets for pH 4.01, 7.00, 9.21 and 10.00, in addition to the meter-only standard accessories.
Spécifications
- Désignation:pH SevenDirect™ SD20 solids kit
- Cat. No.:CA76533-444
- pH accuracy:0.002
- pH resolution:0.1; 0.01; 0.001
- Temperature range:–30...130 °C
- mV accuracy:0.1
- mV resolution:1; 0.1
Accessories
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Parking beaker EasyPlace™Parking beaker EasyPlace™76533-476The parking beaker EasyPlace™ for the SevenDirect™ pH meter is robust and resistant to chemicals for most common lab applications. The beaker has convenient 20 mL graduations for quick and easy measurements.Log in pour la disponibilitéEach - 33,90 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Parking beaker insertParking beaker insert76533-478The parking beaker insert EasyPlace™ for the SevenDirect™ pH meter is secure, keeping the beaker in place while working with various lab samples.Log in pour la disponibilitéEach - 13,20 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Sachet holder EasyPlace™Sachet holder EasyPlace™76533-480The sachet holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sachets in place. The EasyPlace™ sachet holder is made for the newly designed pH and conductivity style sachets to ensure calibration is quick and easy.Log in pour la disponibilitéEach - 69,30 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Sensor holder EasyPlace™ tripleSensor holder EasyPlace™ triple76533-484The sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements.Log in pour la disponibilitéEach - 41,60 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Pole EasyPlace™ 30 cmPole EasyPlace™ 30 cm76533-488The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Log in pour la disponibilitéEach - 339,00 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Pole EasyPlace™ 50 cmPole EasyPlace™ 50 cm76533-490The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Log in pour la disponibilitéEach - 402,00 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Base plate EasyPlace™Base plate EasyPlace™76533-492The base plate EasyPlace™ for the SevenDirect™ meter is extremely secure, distributing weight evenly over the entire meter area. Meter parts such as the electrode arm will be attached to the base plate when you start to build your SevenDirect pH meter.Log in pour la disponibilitéEach - 203,00 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Power supply 18WPower supply 18W76533-494The power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter.Log in pour la disponibilitéEach - 135,00 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Plug set 18 W power supplyPlug set 18 W power supply76533-496The plug set power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter. Additional adaptors included in the plug set.Log in pour la disponibilitéEach - 45,70 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Top cover pole EasyPlace™Top cover pole EasyPlace™76533-498The top cover pole EasyPlace™ fits tightly over the EasyPlace™ stand for an added layer of protection when not operating the meter.Log in pour la disponibilitéEach - 33,90 $
Informations sur le produit
- Catalog #DetailsAvailabilityPrice
- Screws and feet set EasyPlace™Screws and feet set EasyPlace™76533-500The EasyPlace™ screws and feet set are included with the base plate for SevenDirect™ meter assembly.Log in pour la disponibilitéEach - 41,60 $