SevenDirect™ SD23 pH/Conductivity Meter
The pH/conductivity meter SevenDirect™ SD23 provides accurate pH and conductivity measurements for almost any sample across a wide range of applications. The EasyPlace electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
- Intuitive user interface using 7" touchscreen
- On-screen instructions guide through the calibration process
- EasyPlace™ electrode arm ensures precise placement of the sensor
- Feature a closed connector compartment to prevent corrosion
- A replacable protective cover keeps the instrument safe from dust and spills (IP 54 in-use)
- Instruments support GLP compliance by storing key measurement data
The intuitive user interface of the generous 7" touchscreen facilitates the measurement process by avoiding unnecessary steps and focusing on the most important information. The SevenDirect™ has a guided verification process that provides a straightforward way to check system performance with a clear pass/fail indication. The meter has a replaceable protective cover to keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use).
: The meter-only model includes EasyPlace™ electrode holder, protective cover, Get-Started guide and power supply. The kits also include the InLab expert Pro-ISM, conductivity sensor InLab 731-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2×2 conductivity standards (1413 μs/cm and 12.88 ms/cm), in addition to the meter-only standard accessories.
The pure H₂O kit includes the InLab Pure Pro-ISM, conductivity sensor InLab 741-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2 conductivity standards (84 μs/cm).
- Désignation:pH/Conductivity meter SevenDirect™ SD23
- Cat. No.:CA76533-434
- pH accuracy:0.002
- pH resolution:0.1; 0.01; 0.001
- Conductivity range:0.001 μs/cm - 2000 ms/cm
- Temperature range:–30...130 °C
- mV resolution:1; 0.1
Accessories
- DescriptionDetails
- 76533-476Parking beaker EasyPlace™The parking beaker EasyPlace™ for the SevenDirect™ pH meter is robust and resistant to chemicals for most common lab applications. The beaker has convenient 20 mL graduations for quick and easy measurements.Each - $33.90
- DescriptionDetails
- 76533-478Parking beaker insertThe parking beaker insert EasyPlace™ for the SevenDirect™ pH meter is secure, keeping the beaker in place while working with various lab samples.Each - $13.20
- DescriptionDetails
- 76533-480Sachet holder EasyPlace™The sachet holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sachets in place. The EasyPlace™ sachet holder is made for the newly designed pH and conductivity style sachets to ensure calibration is quick and easy.Each - $69.30
- DescriptionDetails
- 76533-488Pole EasyPlace™ 30 cmThe pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Each - $339.00
- DescriptionDetails
- 76533-484Sensor holder EasyPlace™ tripleThe sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements.Each - $41.60
- DescriptionDetails
- 76533-490Pole EasyPlace™ 50 cmThe pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Each - $402.00
- DescriptionDetails
- 76533-492Base plate EasyPlace™The base plate EasyPlace™ for the SevenDirect™ meter is extremely secure, distributing weight evenly over the entire meter area. Meter parts such as the electrode arm will be attached to the base plate when you start to build your SevenDirect pH meter.Each - $203.00
- DescriptionDetails
- 76533-494Power supply 18WThe power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter.Each - $135.00
- DescriptionDetails
- 76533-496Plug set 18 W power supplyThe plug set power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter. Additional adaptors included in the plug set.Each - $45.70
- DescriptionDetails
- 76533-498Top cover pole EasyPlace™The top cover pole EasyPlace™ fits tightly over the EasyPlace™ stand for an added layer of protection when not operating the meter.Each - $33.90
- DescriptionDetails
- 76533-500Screws and feet set EasyPlace™The EasyPlace™ screws and feet set are included with the base plate for SevenDirect™ meter assembly.Each - $41.60