Wheaton Rectangular Staining Dish with Glass Cover | Avantor
Wheaton Rectangular Staining Dish with Glass Cover
10 - 20 Slide staining dish with cover, rack, and handle
Catalog # 25461-002
Fournisseur #:  900200
Wheaton Rectangular Staining Dish with Glass Cover
Catalog # 25461-002
Wheaton Rectangular Staining Dish with Glass Cover
Wheaton Rectangular Staining Dish with Glass Cover
10 - 20 Slide staining dish with cover, rack, and handle
Price
$1,319.50
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 6
Product Details & Documents
The 10 - 20 slide unit is the standard for manual staining procedures.
The removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back. Standard slide sizes accommodated are 3×1" (75×25 mm), 3×11/2" (75×38 mm), and 3×2" (75×51 mm). Manufactured from soda-lime glass. Approximate inside dimensions are 91L×71W×60D mm.
Spécifications
  • Désignation:
    10 - 20 Slide staining dish with cover, rack, and handle
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 10–20 Slide Staining Dish Only
    10–20 Slide Staining Dish Only
    25461-010
    Log in pour la disponibilité
    Case of 3 - $273.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 10–20 Slide Staining Dish Cover Only
    10–20 Slide Staining Dish Cover Only
    25461-012
    Log in pour la disponibilité
    Case of 3 - $172.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stainless Steel Handle
    Stainless Steel Handle
    25464-001
    Log in pour la disponibilité
    Pack of 3 - $55.60
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 10–20 Slide Staining Dish with Cover
    10–20 Slide Staining Dish with Cover
    25462-006
    Log in pour la disponibilité
    Each - $149.24
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 10–20 Slide Glass Rack
    10–20 Slide Glass Rack
    25463-009
    Log in pour la disponibilité
    Each - $87.09