Wheaton Rectangular Staining Dish with Glass Cover
10 - 20 Slide staining dish with cover, rack, and handle
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 900200
Wheaton Rectangular Staining Dish with Glass Cover
Catalog # 25461-002
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
The 10 - 20 slide unit is the standard for manual staining procedures.
The removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back. Standard slide sizes accommodated are 3×1" (75×25 mm), 3×11/2" (75×38 mm), and 3×2" (75×51 mm). Manufactured from soda-lime glass. Approximate inside dimensions are 91L×71W×60D mm.
Documents
Spécifications
- Désignation:10 - 20 Slide staining dish with cover, rack, and handle
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- 10–20 Slide Staining Dish Only
Informations sur le produit
- Catalog #AvailabilityPrice
- 10–20 Slide Staining Dish Cover Only
Informations sur le produit
- Catalog #AvailabilityPrice
- Stainless Steel Handle
Informations sur le produit
- Catalog #AvailabilityPrice
- 10–20 Slide Staining Dish with Cover
Informations sur le produit
- Catalog #AvailabilityPrice
- 10–20 Slide Glass Rack