Control Slides, Normal Human Tissue, Cervix
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
- Product specific reactivity is guaranteed for one year from date of receipt
- Revalidate after one year to verify continued reactivity
- All controls are cut in serial sections at 4 µ
- Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
- All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
: Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
- Désignation:Mopec control slides, normal human tissue, cervix