Control Slides IHC CD 43
Control Slides IHC CD 43
Mopec Control slides IHC CD 43
  76947-730
 :   Mopec
Supplier #:  SC599-100
Control Slides IHC CD 43
  76947-730
 :  Mopec
Control Slides IHC CD 43
Control Slides IHC CD 43
Mopec Control slides IHC CD 43
$2,554.56
Case of 100
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 μm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Ordering information: Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
  • Désignation:
    Mopec Control slides IHC CD 43
  • Cat. No.:
    76947-730