Control Slides IHC SMAD4/DPC4 | Avantor
Control Slides IHC SMAD4/DPC4
Mopec control slides IHC SMAD4/DPC4
Catalog # 76947-398
Marketsource
Fournisseur #:  SC515-25
Control Slides IHC SMAD4/DPC4
Catalog # 76947-398
Supplier:  
Marketsource
Control Slides IHC SMAD4/DPC4
Control Slides IHC SMAD4/DPC4
Mopec control slides IHC SMAD4/DPC4
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
 : Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
Spécifications
  • Désignation:
    Mopec control slides IHC SMAD4/DPC4