Control Slides, Normal Human Tissue, Tonsil | Avantor
Control Slides, Normal Human Tissue, Tonsil
Control slides, normal human tissue, tonsil
Catalog # 76948-128
Supplier:   Mopec
Fournisseur #:  SC922-25
Control Slides, Normal Human Tissue, Tonsil
Catalog # 76948-128
Supplier:  Mopec
Control Slides, Normal Human Tissue, Tonsil
Control Slides, Normal Human Tissue, Tonsil
Control slides, normal human tissue, tonsil
Price
$751.55
Log in pour la disponibilité
Case of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
 : Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
Spécifications
  • Désignation:
    Control slides, normal human tissue, tonsil
  • Cat. No.:
    76948-128