PIP® Dynamic Bump Caps | Avantor
PIP® Dynamic Bump Caps
Bump cap, HDPE, Grey
Catalog # 76944-660
Supplier:   PIP
Fournisseur #:  280-HP940-09
PIP® Dynamic Bump Caps
Catalog # 76944-660
Supplier:  PIP
PIP® Dynamic Bump Caps
PIP® Dynamic Bump Caps
Bump cap, HDPE, Grey
Price
$6.95
Log in pour la disponibilité
Each Case of 20
Product Details & Documents
Bump Caps feature a four-point plastic suspension and pin-lock adjustable back.
  • Lightweight ventilated HDPE shell
  • Four-point HDPE pin-lock suspension
  • With foam sweatband
Popular in food processing, manufacturing, maintenance, and service industries, the bump cap is designed as a safeguard against accidental bumping or scraping of the head.
Compatible face shield bracket 251-EPB940 and Visor 251-FA815. Compatible with chin strap 280-HP241C. Does not meet and is not tested to any safety standards.
Spécifications
  • Color:
    Grey
  • Désignation:
    Bump cap
  • Matière:
    HDPE