PIP® Dynamic Bump Caps
Bump cap, HDPE, White
Catalog # 76944-648
Supplier: PIP
Supplier #: 280-HP940-01
Jump to:
Product Details & Documents
Bump Caps feature a four-point plastic suspension and pin-lock adjustable back.
- Lightweight ventilated HDPE shell
- Four-point HDPE pin-lock suspension
- With foam sweatband
Popular in food processing, manufacturing, maintenance, and service industries, the bump cap is designed as a safeguard against accidental bumping or scraping of the head.
Compatible face shield bracket 251-EPB940 and Visor 251-FA815. Compatible with chin strap 280-HP241C. Does not meet and is not tested to any safety standards.
Specifications
- Color:White
- Désignation:Bump cap
- Matière:HDPE