Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ wipes, flat stacked, Cellulose/polyester, 17.8×17.8 cm (7×7")
Catalog # 76945-154
Jump to:
Discontinued
Product Details & Documents
Amplitude™ Lambda™ Wipes are a hydroentangled blend of 58% cellulose and 42% polyester. The Lambda™ wipe is highly sorbent, has excellent durability, and good cleanliness.
- Hydroentangled fabric provides low levels of particles and extractables
- Excellent for wiping and absorbing spills
- Strong and durable both wet and dry
- Good cost effective, general-purpose wipe
An excellent all-purpose wipe with outstanding sorbency, it is ideal for general-purpose cleaning and spill pick-up in biotech, pharmaceutical, and medical device industries.
Recommended for ISO Class 6 – 8 environments.
Spécifications
- Désignation:Amplitude™ Lambda™ wipes, flat stacked
- Matière:Cellulose/polyester
- W×L:17.8×17.8 cm (7×7")