RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 1 ml glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching silicone rubber, plug style, sealing mat
Catalog # 76881-740
Supplier: MicroSolv Technology Corp
Fournisseur #: 95296-1ML-1PP
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Catalog # 76881-740
Supplier: MicroSolv Technology Corp
Jump to:
Product Details & Documents
Spécifications
- Désignation:Micro-Sample Management System. Complete set, includes 96 individual, 1 ml glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching silicone rubber, plug style, sealing mat