RSA-Pro X™ Micro-Sample Management Systems and Inserts
Inserts, RSA-Pro X™, in racks. Includes 576 clear, 500 µl surface treated glass inserts with wide conical points in 6 Racks of 96
76881-774
: 95296-500-P-PK
- Désignation:Inserts, RSA-Pro X™, in racks. Includes 576 clear, 500 µl surface treated glass inserts with wide conical points in 6 Racks of 96