RSA-Pro X™ Micro-Sample Management Systems and Inserts
Inserts, RSA-Pro X™, in racks. Includes 1,728 clear, 500 µl surface treated glass inserts with wide conical points in 18 Racks of 96
Catalog # 76881-772
Supplier: MicroSolv Technology Corp
Fournisseur #: 95296-500-P-CS
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Catalog # 76881-772
Supplier: MicroSolv Technology Corp
Jump to:
Product Details & Documents
Spécifications
- Désignation:Inserts, RSA-Pro X™, in racks. Includes 1,728 clear, 500 µl surface treated glass inserts with wide conical points in 18 Racks of 96