CELLS MOUSE CCR9 HEK293T ARRESTED 11VL
Catalogue # 76846-880
Fournisseur: MULTISPAN INC
CELLS MOUSE CCR9 HEK293T ARRESTED 11VL
Catalogue # 76846-880
Fournisseur: MULTISPAN INC
Supplier Number: DCM1017-11VL
Frequently Bought Together
Acide chlorhydrique 0.5 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Fournisseur: Air-Tite
Formalin 10%, neutral buffered, VWR®
Fournisseur: VWR International
VWR® Sleeve Stoppers
Fournisseur: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Hemoccult® Guaiac Fecal Occult Blood Test Systems, Beckman Coulter®
Fournisseur: Beckman Coulter
96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Tetrabutylammonium bromide ≥98.0% (by titrimetric analysis)
Fournisseur: TCI America
BBL™ IsoVitaleX™ Enrichment Media, BD Diagnostics
Fournisseur: BD
Ozone AccuVac® Ampules, MR, Hach
Fournisseur: Hach
Calcium carbonate ≥99.0% ACS low in alkali
Fournisseur: Thermo Scientific Chemicals
STORAGE SOLUTION PH ELECTRODE
Fournisseur: Oakton
Forget-Me-Not™ EvaGreen® qPCR Master Mixes, Biotium
Fournisseur: Biotium
Dimethyl sulfoxide ≥99.9% for HPLC
Fournisseur: Thermo Scientific Chemicals
Gibson Assembly Master Mix, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Ammonium acetate ≥97% for HPLC, J.T.Baker®
Fournisseur: VWR International